Merck
  • "Sponge" nanoparticle dispersions in aqueous mixtures of diglycerol monooleate, glycerol dioleate, and polysorbate 80.
"Sponge" nanoparticle dispersions in aqueous mixtures of diglycerol monooleate, glycerol dioleate, and polysorbate 80.
Langmuir : the ACS journal of surfaces and colloids (2006-06-28)
Justas Barauskas, Audrius Misiunas, Torsten Gunnarsson, Fredrik Tiberg, Markus Johnsson
ZUSAMMENFASSUNG
Lipid nanoparticles of nonlamellar lyotropic phases have a wide solubilizing and encapsulating spectrum for a range of substances thanks to their nanostructured interior featuring both lipophilic and hydrophilic domains. As a consequence, these systems have emerged as promising drug delivery systems in various pharmaceutical and diagnostic applications. Here we present the phase behavior and dispersion properties of a novel three-component lipid system composed of diglycerol monooleate (DGMO), glycerol dioleate (GDO), and polysorbate 80 (P80) which shows several advantageous features relating to drug delivery applications including: spontaneous dispersion formation with a narrow size distribution and tunable particle phase-structure. The obtained phase diagram shows the presence of lamellar (L(alpha)), hexagonal (H(2)), and reverse bicontinuous cubic (V(2)) liquid crystalline phases and an inverse micellar (L(2)) solution. A particularly interesting observation is the presence of a phase region where two liquid phases coexist, most likely the L(2) and L(3) ("sponge phase"). These two phase structures appear also to coexist in the submicron particles formed in the dilute water region, where the L(3) element appears to stabilize nanoparticles with inner L(2) structure. Increasing the fraction of the dispersing P80 component results in the growth of the more water rich L(3) "surface phase" at the expense of the size of the inner L(2) core.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Dioleoylglycerin, ≥99%, mixture of 1,3- and 1,2-isomers, liquid