Merck
  • A liquid chromatography/mass spectrometry-based method for the selection of ATP competitive kinase inhibitors.
A liquid chromatography/mass spectrometry-based method for the selection of ATP competitive kinase inhibitors.
Journal of biomolecular screening (2005-08-12)
Sanjay S Khandekar, Bingbing Feng, Tracey Yi, Susan Chen, Nicholas Laping, Neal Bramson
ZUSAMMENFASSUNG
The currently approved kinase inhibitors for therapeutic uses and a number of kinase inhibitors that are undergoing clinical trials are directed toward the adenosine triphosphate (ATP) binding site of protein kinases. The 5'-fluorosulfonylbenzoyl 5'-adenosine (FSBA) is an ATP-affinity reagent that covalently modifies a conserved lysine present in the nucleotide-binding site of most kinases. The authors have developed a liquid chromatography/mass spectrometry-based method to monitor binding of ATP competitive protein kinase inhibitors using FSBA as a nonselective activity-based probe for protein kinases. Their method provides a general, rapid, and reproducible means to screen and validate selective ATP competitive inhibitors of protein kinases.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
5′-(4-Fluorosulfonylbenzoyl)adenosin -hydrochlorid