Merck
  • [Inhibition of respiratory syncytial virus (RSV) by moroxydine, allicin, alpha-interferon and their joint application].
[Inhibition of respiratory syncytial virus (RSV) by moroxydine, allicin, alpha-interferon and their joint application].
Zhonghua shi yan he lin chuang bing du xue za zhi = Zhonghua shiyan he linchuang bingduxue zazhi = Chinese journal of experimental and clinical virology (2004-12-25)
G Zhou, S Han, Y Shi, H Sui
ZUSAMMENFASSUNG
This paper reported the inhibition of respiratory syncytial virus by Moroxydine (ABOB), Allicin, alpha-interferon and their joint application. The results indicated that each of the three drugs inhibited RSV in HeLa cells, and the effective dosage of ABOB was 100 mg/L, alpha-interferon 1.5 x 10(4) U/L and Allicin one eight hundred thousandth of the pure material. The combinations of 3 drugs showed synergism without antagonism, this not only decreased the dosage, toxicity but also enhanced the inhibition effects on respiratory syncytial virus obviously.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Moroxydin -hydrochlorid, 99%