Merck
  • Somatostatin derivative (smsDX) targets cellular metabolism in prostate cancer cells after androgen deprivation therapy.
Somatostatin derivative (smsDX) targets cellular metabolism in prostate cancer cells after androgen deprivation therapy.
PloS one (2013-02-15)
Lei Yan, Zhaoquan Xing, Zhaoxin Guo, Zhiqing Fang, Wei Jiao, Xiaoyu Guo, Zhonghua Xu, Zhenghui Fang, Anders Holmberg, Sten Nilsson, Zhaoxu Liu
ZUSAMMENFASSUNG
Cancer cell metabolism responsive to androgen deprivation therapy (ADT) may be involved in the development and progression of prostate cancer and the ultimate failure of androgen-deprivation therapy. To investigate the metabolism regulation effects on androgen-independent growth of prostate cancer, an established LNCaP-s cell model that resembles the clinical scenario of castration-resistant prostate cancer (CRPC), was used in this current study. This cell line was cultured from androgen-sensitive LNCaP parental cells, in an androgen-reduced condition, resembling clinical androgen deprivation therapy. To assess the effects of smsDX on the invasiveness of prostate cancer cells we used wound healing assay and Matrigel™ invasion assay. We evaluated differentially expressed proteins of the parental LNCaP cells and LNCaP-s cells after ADT by means of two-dimensional gel electrophoresis (2-DE) followed by MALDI-TOF mass spectrometric analysis. The covered area in the wound and the number of cells invading through a Matrigel chamber were significantly smaller for cells treated with smsDX than they were for control cells treated with vehicle. 56 proteins were found differentially expressed in LNCaP-s cells compared to LNCaP cells, majority of them were down-regulated after ADT treatment. 104 proteins of LNCaP cells and 86 in LNCaP-s cells, separately, were found differentially expressed after treatment with smsDX, When we explored these protein functions within the website UniProtKB/Swiss-Prot, surprisingly, most of the proteins were found to be involved in the cellular metabolism and mitochondrial function regulation. LNCaP-s as potential metastatic androgen-independent cancer cells, its metabolism and mitochondrial functions could be altered by a new somatostatin derivative smsDX, the smsDX regulatory effects on metabolism in LNCaP-s deliver more therapeutic information with the treatment of CRPC.