Merck
Desmoplastic stromal signatures predict patient outcomes in pancreatic ductal adenocarcinoma.
Cell reports. Medicine (2023-10-22)
Shamik Mascharak, Jason L Guo, Deshka S Foster, Anum Khan, Michael F Davitt, Alan T Nguyen, Austin R Burcham, Malini S Chinta, Nicholas J Guardino, Michelle Griffin, David M Lopez, Elisabeth Miller, Michael Januszyk, Shyam S Raghavan, Teri A Longacre, Daniel J Delitto, Jeffrey A Norton, Michael T Longaker
ZUSAMMENFASSUNG
Pancreatic ductal adenocarcinoma (PDAC) is projected to become the second leading cause of cancer-related death. Hallmarks include desmoplasia with variable extracellular matrix (ECM) architecture and a complex microenvironment with spatially defined tumor, stromal, and immune populations. Nevertheless, the role of desmoplastic spatial organization in patient/tumor variability remains underexplored, which we elucidate using two technologies. First, we quantify ECM patterning in 437 patients, revealing architectures associated with disease-free and overall survival. Second, we spatially profile the cellular milieu of 78 specimens using codetection by indexing, identifying an axis of pro-inflammatory cell interactions predictive of poorer outcomes. We discover that clinical characteristics, including neoadjuvant chemotherapy status, tumor stage, and ECM architecture, correlate with differential stromal-immune organization, including fibroblast subtypes with distinct niches. Lastly, we define unified signatures that predict survival with areas under the receiver operating characteristic curve (AUCs) of 0.872-0.903, differentiating survivorship by 655 days. Overall, our findings establish matrix ultrastructural and cellular organizations of fibrosis linked to poorer outcomes.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Anti-FAP (AB1) antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution