- Analysis of microglial production of reactive oxygen and nitrogen species.
Analysis of microglial production of reactive oxygen and nitrogen species.
Methods in molecular biology (Clifton, N.J.) (2013-07-03)
Urte Neniskyte, Guy C Brown
PMID23813374
ZUSAMMENFASSUNG
Reactive oxygen and nitrogen species are both regulators and effectors of microglial activation, and assays of these oxidants can be used as a measure of acute and chronic activation of microglial cells. Here we describe quick methods to assess the production of superoxide, hydrogen peroxide, nitric oxide, and peroxynitrite by microglia.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Katalase aus Humanerythrocyten, ≥90% (SDS-PAGE), buffered aqueous solution, ≥30,000 units/mg protein