Merck
  • Paraoxonase-1 suppresses experimental colitis via the inhibition of IFN-γ production from CD4 T cells.
Paraoxonase-1 suppresses experimental colitis via the inhibition of IFN-γ production from CD4 T cells.
Journal of immunology (Baltimore, Md. : 1950) (2013-06-19)
Junji Yamashita, Chiaki Iwamura, Toshihiro Ito, Masakuni Narita, Yukio Hara, Tetsuya Sasaki, Daisuke Masuda, Munehisa Takahashi, Manami Tsuchiya, Kaori Hada, Makoto Ishikawa, Takato Matsuo, Yoichi Ohno, Hitoshi Tanaka, Hideya Maruyama, Yasumasa Ogawa, Toshinori Nakayama
ZUSAMMENFASSUNG
Crohn's disease (CD) is a chronic inflammatory disorder of the gastrointestinal tract, where excessive Th1 cell responses are observed. We performed experiments to identify immunologically bioactive proteins in human plasma and found that paraoxonase (PON)-1, which has esterase activity and is associated with high-density lipoproteins, inhibited the IFN-γ production by both murine and human differentiating Th1 cells. Trinitrobenzene sulfonic acid-induced colitis was attenuated by the administration of PON-1. The beneficial effects of PON-1 were associated with a reduced ratio of IFN-γ-producing CD4 T cells in the mesenteric lymph nodes and decreased production of T cell-related cytokines in the colon. PON-1 inhibited the TCR-induced activation of ERK-MAPK signaling and the nuclear translocation of NF-κB in CD4 T cells. Interestingly, an excessive CD4 T cell response was observed in PON-1-deficient mice under physiological and pathological conditions. Additionally, the efficacy of PON-1 or G3C9-C284A (G3C9), which shows a higher esterase activity than PON-1, on colitis was similar to that of an anti-TNF-α mAb, which is a clinically used CD treatment. Moreover, G3C9 more effectively suppressed CD4(+)CD45RB(high) cell transfer-induced chronic colitis in mice than did PON-1, and the efficacy of G3C9 against the colitis was similar to that of the anti-TNF-α mAb. Therefore, PON-1 (or G3C9) administration may be clinically beneficial for CD patients.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
SAFC
CHOZN® CHO K1 Host Cell Line, Suspension-adapted in CD media