- Helix-helix interfaces and ligand binding.
Helix-helix interfaces and ligand binding.
Journal of theoretical biology (2011-05-31)
Natalya Kurochkina, Tsering Choekyi
PMID21620863
ZUSAMMENFASSUNG
Helix-helix parallel interfaces can be characterized by certain combinations of amino acids, which repeatedly occur at core positions a and d (leucine zipper nomenclature) in homologous and nonhomologous proteins and influence interhelical angles. Applied for the prediction of interhelical angles in glutathione S-transferase, intracellular chloride channel and annexin molecules from various sources, correct results were achieved in 58 out of 62 proteins. Interhelical angles are found to correlate with the conformation of the glutathione S-transferase ligands glutathione, s-hexylglutathione, glutathione sulfonic acid, and glutathione-s-dinitrobenzene.
MATERIALIEN