Merck
  • Antiamyloidogenic and neuroprotective functions of cathepsin B: implications for Alzheimer's disease.
Antiamyloidogenic and neuroprotective functions of cathepsin B: implications for Alzheimer's disease.
Neuron (2006-09-20)
Sarah Mueller-Steiner, Yungui Zhou, Hideaki Arai, Erik D Roberson, Binggui Sun, Jennifer Chen, Xin Wang, Guiqiu Yu, Luke Esposito, Lennart Mucke, Li Gan
ZUSAMMENFASSUNG
Alzheimer's disease (AD) may result from the accumulation of amyloid-beta (Abeta) peptides in the brain. The cysteine protease cathepsin B (CatB) is associated with amyloid plaques in AD brains and has been suspected to increase Abeta production. Here, we demonstrate that CatB actually reduces levels of Abeta peptides, especially the aggregation-prone species Abeta1-42, through proteolytic cleavage. Genetic inactivation of CatB in mice with neuronal expression of familial AD-mutant human amyloid precursor protein (hAPP) increased the relative abundance of Abeta1-42, worsening plaque deposition and other AD-related pathologies. Lentivirus-mediated expression of CatB in aged hAPP mice reduced preexisting amyloid deposits, even thioflavin S-positive plaques. Under cell-free conditions, CatB effectively cleaved Abeta1-42, generating C-terminally truncated Abeta peptides that are less amyloidogenic. Thus, CatB likely fulfills antiamyloidogenic and neuroprotective functions. Insufficient CatB activity might promote AD; increasing CatB activity could counteract the neuropathology of this disease.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Cathepsin B from human placenta, lyophilized powder, ≥5 units/mg protein