Merck
  • Directly Microwave-Accelerated Cleavage of C-C and C-O Bonds of Lignin by Copper Oxide and H2 O2.
Directly Microwave-Accelerated Cleavage of C-C and C-O Bonds of Lignin by Copper Oxide and H2 O2.
ChemSusChem (2020-04-11)
Chen Qu, Keigo Ito, Isamu Katsuyama, Tomohiko Mitani, Keiichiro Kashimura, Takashi Watanabe
ZUSAMMENFASSUNG
Model erythro, phenolic, and nonphenolic lignin β-O-4 dimer compounds are treated with copper oxide and H2 O2 at the electronic field maximum position of a single-mode 2.45 GHz microwave system equipped with a cavity resonator. The products obtained through microwave heating and oil-bath heating with the same reaction vessel and temperature profile are quantitatively compared. Dimer degradation is found to proceed through consecutive elementary reactions. The phenolic dimer is dehydroxylated and this is followed by the spontaneous cleavage of Cα -Cβ and C-O-C bonds to produce guaiacol, vanillin, and vanillic acid. The reaction of the nonphenolic dimer produces veratric acid, veratraldehyde, and guaiacol. Microwave irradiation accelerates cleavage of the side chain and the oxidation of vanillin to vanillic acid. However, no acceleration of veratraldehyde oxidation to veratric acid or aromatic ring cleavage to produce dicarboxylic acids is observed. The selective acceleration of elementary reactions during the degradation of model lignin compounds indicates that microwaves interact with reaction intermediates that are sensitive to electromagnetic waves.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
guaiacylglycerol-beta-guaiacyl ether, AldrichCPR