Merck
  • A Phase II Study of Cabozantinib and Androgen Ablation in Patients with Hormone-Naïve Metastatic Prostate Cancer.
A Phase II Study of Cabozantinib and Androgen Ablation in Patients with Hormone-Naïve Metastatic Prostate Cancer.
Clinical cancer research : an official journal of the American Association for Cancer Research (2020-01-17)
Paul G Corn, Miao Zhang, Graciela M Nogueras-Gonzalez, Lianchun Xiao, Amado J Zurita, Sumit K Subudhi, Shi-Ming Tu, Ana M Aparicio, Cristian Coarfa, Kimal Rajapakshe, Shixia Huang, Nora M Navone, Sue-Hwa Lin, Guocan Wang, Sumankalai Ramachandran, Mark A Titus, Theocharis Panaretakis, Gary E Gallick, Eleni Efstathiou, Patricia Troncoso, Christopher Logothetis
ZUSAMMENFASSUNG
Cabozantinib, an oral inhibitor of c-MET/VEGFR2 signaling, improved progression-free survival (mPFS) but not overall survival (OS) in metastatic castrate-resistant prostate cancer. We evaluated cabozantinib plus androgen deprivation therapy (ADT) in hormone-naïve metastatic prostate cancer (HNMPCa). Patients received ADT plus cabozantinib starting at 60 mg daily. The primary endpoint was castrate-resistant PFS by radiographic criteria, clinical progression, or receipt of additional therapy. Secondary endpoints included OS, safety, radiographic responses, and biomarker modulation. Sixty-two patients received treatment. With a median follow-up of 31.2 months, the mPFS was 16.1 months (95% CI, 14.6-22.7 months), and mOS was not reached. Reductions in PSA ≥ 90%, bone-specific alkaline phosphatase ≥ 50%, and urine N-telopeptides ≥ 50% occurred in 83%, 87%, and 86% of evaluable patients, respectively. Responses in bone scan and measurable disease were observed in 81% of and 90% of evaluable patients, respectively. Most common grade 3 adverse events were hypertension (19%), diarrhea (6%), and thromboembolic events (6%), and dose reductions occurred in 85% of patients. Analysis of baseline cytokine and angiogenic factors (CAFs) revealed that higher plasma concentrations of Lumican, CXCL5, CD25, and CD30 were associated with shorter PFS as was high tumor expression of pFGFR1. Cabozantinib plus ADT has promising clinical activity in HNMPCa. CAF profiles and tissue markers suggest candidate prognostic and predictive markers of cabozantinib benefit and provide insights for rational therapy combinations.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Millipore
MILLIPLEX® Human Cancer/ Metastasis Biomarker Magnetic Bead Panel - Cancer Multiplex Assay, Circulating Cancer and Metastasis Biomarker Bead-Based Multiplex Assays using the Luminex technology enables the simultaneous analysis of multiple oncology biomarkers in various tumor types in human serum, plasma and cell culture samples.