Merck
  • Inhibition of acetylcholinesterase, beta-amyloid aggregation, and NMDA receptors in Alzheimer's disease: a promising direction for the multi-target-directed ligands gold rush.
Inhibition of acetylcholinesterase, beta-amyloid aggregation, and NMDA receptors in Alzheimer's disease: a promising direction for the multi-target-directed ligands gold rush.
Journal of medicinal chemistry (2008-07-09)
Michela Rosini, Elena Simoni, Manuela Bartolini, Andrea Cavalli, Luisa Ceccarini, Nicoleta Pascu, David W McClymont, Andrea Tarozzi, Maria L Bolognesi, Anna Minarini, Vincenzo Tumiatti, Vincenza Andrisano, Ian R Mellor, Carlo Melchiorre
ZUSAMMENFASSUNG
Alzheimer's disease (AD) is a multifactorial syndrome with several target proteins contributing to its etiology. To confront AD, an innovative strategy is to design single chemical entities able to simultaneously modulate more than one target. Here, we present compounds that inhibit acetylcholinesterase and NMDA receptor activity. Furthermore, these compounds inhibit AChE-induced Abeta aggregation and display antioxidant properties, emerging as lead candidates for treating AD.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
4-Hydroxycarbazol, 95%