Merck
Analysis of base content in in-service oils by fourier transform infrared spectroscopy.
Journal of laboratory automation (2012-02-24)
Sadia Ehsan, Jacqueline Sedman, Frederick R van de Voort, Emmanuel Akochi-Koblé, Tao Yuan, Djaouida Takouk
ZUSAMMENFASSUNG
An automated FTIR method for the determination of the base content (BC(pKa)) of oils at rates of > 120 samples/h has been developed. The method uses a 5% solution of trifluoroacetic acid in 1-propanol (TFA/P) added to heptane-diluted oil to react with the base present and measures the ν(COO(-)) absorption of the TFA anion produced, with calibrations devised by gravimetrically adding 1-methylimidazole to a heptane-TFA/P mixture. To minimize spectral interferences, all spectra are transformed to 2(nd) derivative spectra using a gap-segment algorithm. Any solvent displacement effects resulting from sample miscibility are spectrally accounted for by measurement of the changes in the 1-propanol overtone band at 1936 cm(-1). A variety of oils were analyzed for BC(0.5), expressed as mEq base/g oil as well as converted to base number (BN) units (mg KOH/g oil) to facilitate direct comparison with ASTM D2896 and ASTM D974 results for the same samples. Linear relationships were obtained between FTIR and D2896 and D974, with the ASTM methods producing higher BN values by factors of ~1.5 and ~1.3, respectively. Thus, the FTIR BC method correlates well with ASTM potentiometric procedures and, with its much higher throughput, promises to be a useful alternative means of rapidly determining reserve alkalinity in commercial oil condition monitoring laboratories.