Merck
  • Parental Dietary Protein Source and the Role of CMKLR1 in Determining the Severity of Dahl Salt-Sensitive Hypertension.
Parental Dietary Protein Source and the Role of CMKLR1 in Determining the Severity of Dahl Salt-Sensitive Hypertension.
Hypertension (Dallas, Tex. : 1979) (2019-01-01)
Justine M Abais-Battad, Hayley Lund, Daniel J Fehrenbach, John Henry Dasinger, Ammar J Alsheikh, David L Mattson
ZUSAMMENFASSUNG
Studies from our laboratory have revealed an important role for the maternal diet and the dietary protein source in the development of hypertension and renal injury in Dahl salt-sensitive (SS) rats. The current study sought to compare salt-induced hypertension, renal damage, and immune cell infiltration in the offspring of breeders fed either a casein- or gluten-based diet, with the hypothesis that offspring from gluten-fed breeders would fail to develop these SS phenotypes. When fed identical diets post-weaning, the F1 generation gluten offspring demonstrated lower mean arterial pressure (149.1±3.1 versus 162.5±5.8 mm Hg), albuminuria (166.2±34.6 versus 250.9±27.8 mg/day), and outer medullary protein casting (7.4±0.8% versus 13.1±1.3%) in response to high salt compared with the casein offspring (n=9-11). The gluten offspring also had fewer CD45+ leukocytes, CD11b/c+ monocytes/macrophages, CD3+ T cells, and CD45R+ B cells infiltrating the kidney. Analysis of the F2 generation gluten offspring also exhibited lower mean arterial pressure and renal damage compared with rats born from casein breeders (n=7-9), with no difference in renal immune cell infiltration. CMKLR1-receptor for the novel prohypertensive adipokine chemerin-was found via polymerase chain reaction array to be significantly upregulated (2.99-fold) in renal T cells isolated from F2 offspring of casein-fed versus gluten-fed parents. Furthermore, CMKLR1 inhibition via α-NETA (2-[α-naphthoyl] ethyltrimethylammonium iodide) treatment significantly attenuated renal immune cell infiltration, hypertension, and renal damage in SS rats fed high salt. Together, these data demonstrate the influence of the parental diet in determining the salt-induced hypertension, renal damage, and inflammatory phenotype of the offspring.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
alpha-NETA, ≥98% (HPLC)