Merck
  • A review of the role of apolipoprotein C-II in lipoprotein metabolism and cardiovascular disease.
A review of the role of apolipoprotein C-II in lipoprotein metabolism and cardiovascular disease.
Metabolism: clinical and experimental (2012-02-07)
Anastazia A Kei, Theodosios D Filippatos, Vasilios Tsimihodimos, Moses S Elisaf
ZUSAMMENFASSUNG
The focus of this review is on the role of apolipoprotein C-II (apoC-II) in lipoprotein metabolism and the potential effects on the risk of cardiovascular disease (CVD). We searched PubMed/Scopus for articles regarding apoC-II and its role in lipoprotein metabolism and the risk of CVD. Apolipoprotein C-II is a constituent of chylomicrons, very low-density lipoprotein, low-density lipoprotein, and high-density lipoprotein (HDL). Apolipoprotein C-II contains 3 amphipathic α-helices. The lipid-binding domain of apoC-II is located in the N-terminal, whereas the C-terminal helix of apoC-II is responsible for the interaction with lipoprotein lipase (LPL). At intermediate concentrations (approximately 4 mg/dL) and in normolipidemic subjects, apoC-II activates LPL. In contrast, both an excess and a deficiency of apoC-II are associated with reduced LPL activity and hypertriglyceridemia. Furthermore, excess apoC-II has been associated with increased triglyceride-rich particles and alterations in HDL particle distribution, factors that may increase the risk of CVD. However, there is not enough current evidence to clarify whether increased apoC-II causes hypertriglyceridemia or is an epiphenomenon reflecting hypertriglyceridemia. A number of pharmaceutical interventions, including statins, fibrates, ezetimibe, nicotinic acid, and orlistat, have been shown to reduce the increased apoC-II concentrations. An excess of apoC-II is associated with increased triglyceride-rich particles and alterations in HDL particle distribution. However, prospective trials are needed to assess if apoC-II is a CVD marker or a risk factor in high-risk patients.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Apolipoprotein C-II from human plasma, ≥95% (SDS-PAGE)