Merck
  • Reactions of 1,2,4-Oxadiazole[4,5-a]piridinium Salts with Alcohols: the Synthesis of Alkoxybutadienyl 1,2,4-Oxadiazoles.
Reactions of 1,2,4-Oxadiazole[4,5-a]piridinium Salts with Alcohols: the Synthesis of Alkoxybutadienyl 1,2,4-Oxadiazoles.
ChemistryOpen (2020-02-07)
Mattia Moiola, Marco Leusciatti, Paolo Quadrelli
ZUSAMMENFASSUNG
1,2,4-Oxadiazole[4,5-a]piridinium salts add alcohols and alkoxides to undergo electrocyclic ring opening affording alkoxybutadienyl 1,2,4-oxadiazole derivatives. The pyridinium salts represent a special class of Zincke salts that are prone to rearrange to give alkoxybutadienyl 1,2,4-oxadiazoles when treated with suitable nucleophiles or, alternatively, to give pyridones in the presence of bicarbonate. The pivotal tuning of the experimental conditions leads to a straightforward synthesis of valuable 1,2,4-oxadiazole derivatives. The mechanism is also discussed in the light of previous observations.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
2-Chloropyridin, 99%