Merck
  • Freeze-dried mucoadhesive polymeric system containing pegylated lipoplexes: Towards a vaginal sustained released system for siRNA.
Freeze-dried mucoadhesive polymeric system containing pegylated lipoplexes: Towards a vaginal sustained released system for siRNA.
Journal of controlled release : official journal of the Controlled Release Society (2016-06-23)
Tania Furst, George R Dakwar, Elisa Zagato, Anna Lechanteur, Katrien Remaut, Brigitte Evrard, Kevin Braeckmans, Geraldine Piel
ZUSAMMENFASSUNG
Topical vaginal sustained delivery of siRNA presents a significant challenge due to the short residence time of formulations. Therefore, a drug delivery system capable to adhere to the vaginal mucosa is desirable, as it could allow a prolonged delivery and increase the effectiveness of the therapy. The aim of this project is to develop a polymeric solid mucoadhesive system, loaded with lipoplexes, able to be progressively rehydrated by the vaginal fluids to form a hydrogel and to deliver siRNA to vaginal tissues. To minimize adhesive interactions with vaginal mucus components, lipoplexes were coated with different derivatives of polyethylene glycol: DPSE-PEG2000, DPSE-PEG750 and ceramide-PEG2000. Based on stability and diffusion properties in simulated vaginal fluids, lipoplexes containing DSPE-PEG2000 were selected and incorporated in hydroxyethyl cellulose (HEC) hydrogels. Solid systems, called sponges, were then obtained by freeze-drying. Sponges meet acceptable mechanical characteristics and their hardness, deformability and mucoadhesive properties are not influenced by the presence of lipoplexes. Finally, mobility and stability of lipoplexes inside sponges rehydrated with vaginal mucus, mimicking in situ conditions, were evaluated by advanced fluorescence microscopy. The release rate was found to be influenced by the HEC concentration and consequently by the viscosity after rehydration. This study demonstrates the feasibility of entrapping pegylated lipoplexes into a solid matrix system for a prolonged delivery of siRNA into the vagina.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Avanti
18:0 PEG750 PE, Avanti Research - A Croda Brand 880620P, powder