Merck
Visible-Light-Induced Alkoxyl Radicals Enable α-C(sp3)-H Bond Allylation.
iScience (2019-12-31)
Jing Zhang, Dan Liu, Song Liu, Yuanyuan Ge, Yu Lan, Yiyun Chen
ZUSAMMENFASSUNG
The alkoxyl radical is an essential reactive intermediate in mechanistic studies and organic synthesis with hydrogen atom transfer (HAT) reactivity. However, compared with intramolecular 1,5-HAT or intermolecular HAT of alkoxyl radicals, the intramolecular 1,2-HAT reactivity has been limited to theoretical studies and rarely synthetically utilized. Here we report the first selective 1,2-HAT of alkoxyl radicals for α-C(sp3)-H bond allylation of α-carbonyl, α-cyano, α-trifluoromethyl, and benzylic N-alkoxylphthalimides. The mechanistic probing experiments, electron paramagnetic resonance (EPR) studies, and density functional theory (DFT) calculations confirmed the 1,2-HAT reactivity of alkoxyl radicals, and the use of protic solvents lowered the activation energy by up to 10.4 kcal/mol to facilitate the α-C(sp3)-H allylation reaction.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Diethyl-1,4-dihydro-2,6-dimethyl-3,5-pyridindicarboxylat, 95%