Merck
MreB actin-mediated segregation of a specific region of a bacterial chromosome.
Cell (2005-02-15)
Zemer Gitai, Natalie Anne Dye, Ann Reisenauer, Masaaki Wachi, Lucy Shapiro
ZUSAMMENFASSUNG
Faithful chromosome segregation is an essential component of cell division in all organisms. The eukaryotic mitotic machinery uses the cytoskeleton to move specific chromosomal regions. To investigate the potential role of the actin-like MreB protein in bacterial chromosome segregation, we first demonstrate that MreB is the direct target of the small molecule A22. We then demonstrate that A22 completely blocks the movement of newly replicated loci near the origin of replication but has no qualitative or quantitative effect on the segregation of other loci if added after origin segregation. MreB selectively interacts, directly or indirectly, with origin-proximal regions of the chromosome, arguing that the origin-proximal region segregates via an MreB-dependent mechanism not used by the rest of the chromosome.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
MreB-störende Verbindung A22, A cell-permeable isothiourea compound that specifically, rapidly and reversibly perturbs MreB function without affecting eukaryotic actin polymerization.