Merck
  • Altered signature of apoptotic endothelial cell-derived microvesicles predicts chronic heart failure phenotypes.
Altered signature of apoptotic endothelial cell-derived microvesicles predicts chronic heart failure phenotypes.
Biomarkers in medicine (2019-06-04)
Alexander E Berezin, Alexander A Kremzer, Tatyana A Samura, Tatyana A Berezina
ZUSAMMENFASSUNG
Aim: to evaluate the associations between signatures of apoptotic endothelial cell-derived microvesicles (MVs) with phenotypes of chronic heart failure (HF). Methods: The study cohort consisted of 388 prospectively involved subjects with HF patients with predominantly reduced left ventricular ejection fraction (HFrEF), HF with preserved ejection fraction (HFpEF) and HF with mid-range ejection fraction (HFmrEF). All biomarkers were measured at baseline. Results: The number of circulating CD31+/annexin V+ MVs in HFrEF and HFmrEF patients was similar. The number of circulating CD144+/annexin V+ MVs in HFrEF patients was significantly higher than HFmrEF and HFpEF. We determined that a combination of number of circulating CD31+/annexin V+ MVs and Gal-3 was the best predictor of HFpEF and that number of circulating CD144+/annexin V+ MVs is able to increase predictive capabilities of soluble ST2 (sST2) and Gal-3 for HFrEF. Conclusion: We found that the number of circulating CD31+/annexin V+ MVs may improve a predictive capacity for conventional HF biomarkers.