Merck
  • Development of a dual-functional conjugate of antigenic peptide and Fc-III mimetics (DCAF) for targeted antibody blocking.
Development of a dual-functional conjugate of antigenic peptide and Fc-III mimetics (DCAF) for targeted antibody blocking.
Chemical science (2019-04-19)
Lin Zhang, Hao Shen, Yiyi Gong, Xiaojing Pang, Meiqi Yi, Lin Guo, Jin Li, Sam Arroyo, Xin Lu, Sergey Ovchinnikov, Gong Cheng, Xudong Liu, Xu Jiang, Shan Feng, Haiteng Deng
ZUSAMMENFASSUNG
Targeted antibody blocking enables characterization of binding sites on immunoglobulin G (IgG), and can efficiently eliminate harmful antibodies from organisms. In this report, we present a novel peptide-denoted as a dual-functional conjugate of antigenic peptide and Fc-III mimetics (DCAF)-for targeted blocking of antibodies. Synthesis of DCAF was achieved by native chemical ligation, and the molecule consists of three functional parts: a specific antigenic peptide, a linker and the Fc-III mimetic peptide, which has a high affinity toward the Fc region of IgG molecules. We demonstrate that DCAF binds the cognate antibody with high selectivity by simultaneously binding to the Fab and Fc regions of IgG. Animal experiments revealed that DCAF molecules diminish the antibody-dependent enhancement effect in a dengue virus infection model, and rescue the acetylcholine receptor by inhibiting the complement cascade in a myasthenia gravis model. These results suggest that DCAFs could have utility in the development of new therapeutics against harmful antibodies.