Merck
  • Interactions among Fungal Community, Fusarium Mycotoxins, and Components of Harvested Wheat under Simulated Storage Conditions.
Interactions among Fungal Community, Fusarium Mycotoxins, and Components of Harvested Wheat under Simulated Storage Conditions.
Journal of agricultural and food chemistry (2019-06-28)
Yingyue Zhang, Fei Pei, Yong Fang, Peng Li, Ji Xia, Lei Sun, Yanyu Zou, Fei Shen, Qiuhui Hu
ZUSAMMENFASSUNG
Economic loss of postharvest wheat under poor storage conditions due to fungal spoilage and mycotoxin contamination is severe. In order to study the influencing factors of the aggravation of mildew in natural wheat during storage, we assessed changes in Fusarium mycotoxins by high performance liquid chromatography, changes in fungal communities by high-throughput sequencing, and changes in biochemical components in wheat stored under artificial simulation conditions. Deoxynivalenol was the dominant Fusarium mycotoxin, reaching 1103 μg/kg at 25 °C with 75% relative humidity after 30 weeks. Under these conditions, Fusarium dominated the fungal communities, and Fusarium graminearum was significantly negatively correlated with glutenin (p < 0.05). Low storage temperatures and low humidity result in lower levels of Fusarium mycotoxins. Different fungi tended to consume different wheat components, and the interaction between environmental and biological factors eventually leads to the deterioration of wheat quality. These findings might provide valuable information for control strategies of mildew occurrence during grain storage.