Merck
  • Dissipation of the Insecticide Cyantraniliprole and Its Metabolite IN-J9Z38 in Proso Millet during Cultivation.
Dissipation of the Insecticide Cyantraniliprole and Its Metabolite IN-J9Z38 in Proso Millet during Cultivation.
Scientific reports (2019-08-14)
Jonghwa Lee, Min Woo Jung, Junghak Lee, Jiho Lee, Yongho Shin, Jeong-Han Kim
ZUSAMMENFASSUNG
The dissipation patterns of cyantraniliprole and its metabolite IN-J9Z38 were investigated using proso millet (Panicum miliaceum) under open-field conditions to establish a pre-harvest interval. A simple and sensitive analytical method was developed for analyzing residues using ultra-high performance liquid chromatography coupled with tandem mass spectrometry (UHPLC-MS/MS) for multiple reaction monitoring of target compounds. The analytical method was validated in terms of the instrumental limit of quantitation, method limit of quantitation, linearity, accuracy, and precision. The method was successfully applied to the analysis of cyantraniliprole and IN-J9Z38 residues in the field samples of four plots, which were treated twice with an oil dispersion formulation, according to the date of pesticide treatment before harvest. In the case of cyantraniliprole in grain and straw, there was a 91.1 and 89.1% decrease, respectively, from the initial residues (14-7 days) to the final plot (40-30 days before harvest). However, IN-J9Z38 gradually increased over time, indicating that cyantraniliprole transformed into IN-J9Z38 during cultivation. The biological half-lives of total cyantraniliprole were 11.3 and 9.4 days for grain and straw, respectively. The results obtained in this study will inform regulation and management of pesticide use for the minor crop proso millet.