Merck
  • In vitro effectiveness of recombinant human lactoferrin and its hydrolysate in alleviating LPS-induced inflammatory response.
In vitro effectiveness of recombinant human lactoferrin and its hydrolysate in alleviating LPS-induced inflammatory response.
Food research international (Ottawa, Ont.) (2019-03-23)
Esmat Aly, Rubén López-Nicolás, Aliaa Ali Darwish, Gaspar Ros-Berruezo, Carmen Frontela-Saseta
ZUSAMMENFASSUNG
This study aimed to evaluate the potential anti-inflammatory role of the most produced form of lactoferrin expressed in various expression systems (Fe-saturated recombinant human Lf, rhLf) and its hydrolysate in concentrations resembles that found in mature human milk. Co-culture model consisted of CaCo-2 and RAW 246.7 cell lines was used to evaluate the potential anti-inflammatory activity of rhLf and its hydrolysate. During this experiment, CaCo-2 monolayer permeability and integrity was assayed through the measurement of transepithelial electrical resistance (TEER values). Also, the production of reactive oxygen species (ROS), nitric oxide (NO) and different cytokines (IL-8, IL-1β, IL-6, IL-10, IL-12p70, and TNF-α) were measured. The treatment with rhLf and its hydrolysate protected the monolayer integrity against LPS effect and reduced IL-8 and ROS production. This effect was dependent on the dose and 2mgmL-1 of rhLf hydrolysate was more effective. The addition of rhLf and its hydrolysate to infant formula is a prominent step towards improving both infant formula functionality and newborn health. Thus, these functional ingredients could be incorporated in infant foods. In this context, ongoing researches are conducted to clarify this effect whether by using synthetic peptides or by using LPS-sepsis animal.