Merck
  • Gel- and Solid-State-Structure of Dialanine and Diphenylalanine Amphiphiles: Importance of C⋅⋅⋅H Interactions in Gelation.
Gel- and Solid-State-Structure of Dialanine and Diphenylalanine Amphiphiles: Importance of C⋅⋅⋅H Interactions in Gelation.
Chemphyschem : a European journal of chemical physics and physical chemistry (2019-02-21)
Abbas D Farahani, Adam D Martin, Hasti Iranmanesh, Mohan M Bhadbhade, Jonathon E Beves, Pall Thordarson
ZUSAMMENFASSUNG
To investigate the role of the capping group in the solution and solid-state self-assembly of short peptide amphiphiles, dialanine and diphenylalanine have been linked via the N-terminus to a benzene (phenyl) and 3-naphthyl capping groups using three different methylene linkers; (CH2 )n , n=0-4 for the benezene and 0, 1 and 2 for the naphthalene capping group. Atomic force microscopy (AFM), oscillatory rheology, circular dichroism (CD), and IR analysis have been employed to understand the properties of these peptide-based hydrogels. Several X-ray structures of these short peptide gelators give useful conformational information regarding packing. A comparison of these solid state structures with their gel state properties yielded greater insights into the process of self-assembly in short peptide gelators, particularly in terms of the important role of C⋅⋅⋅H interactions appear to play in determining if a short aromatic peptide does form a gel or not.