Merck
  • Preclinical assessment of the ADME, efficacy and drug-drug interaction potential of a novel NAMPT inhibitor.
Preclinical assessment of the ADME, efficacy and drug-drug interaction potential of a novel NAMPT inhibitor.
Xenobiotica; the fate of foreign compounds in biological systems (2018-09-28)
Bianca M Liederer, Jonathan Cheong, Kang-Jye Chou, Peter S Dragovich, Hoa Le, Xiaorong Liang, Justin Ly, Sophie Mukadam, Jason Oeh, Deepak Sampath, Leslie Wang, Susan Wong
ZUSAMMENFASSUNG
GNE-617 (N-(4-((3,5-difluorophenyl)sulfonyl)benzyl)imidazo[1,2-a]pyridine-6-carboxamide) is a potent, selective nicotinamide phosphoribosyltransferase (NAMPT) inhibitor being explored as a potential treatment for human cancers. Plasma clearance was low in monkeys and dogs (9.14 mL min-1 kg-1 and 4.62 mL min-1 kg-1, respectively) and moderate in mice and rats (36.4 mL min-1 kg-1 and 19.3 mL min-1 kg-1, respectively). Oral bioavailability in mice, rats, monkeys and dogs was 29.7, 33.9, 29.4 and 65.2%, respectively. Allometric scaling predicted a low clearance of 3.3 mL min-1 kg-1 and a volume of distribution of 1.3 L kg-1 in human. Efficacy (57% tumor growth inhibition) in Colo-205 CRC tumor xenograft mice was observed at an oral dose of 15 mg/kg BID (AUC = 10.4 µM h). Plasma protein binding was moderately high. GNE-617 was stable to moderately stable in vitro. Main human metabolites identified in human hepatocytes were formed primarily by CYP3A4/5. Transporter studies suggested that GNE-617 is likely a substrate for MDR1 but not for BCRP. Simcyp® simulations suggested a low (CYP2C9 and CYP2C8) or moderate (CYP3A4/5) potential for drug-drug interactions. The potential for autoinhibition was low. Overall, GNE-617 exhibited acceptable preclinical properties and projected human PK and dose estimates.