Merck
  • Interleukin 6-triggered ataxia-telangiectasia mutated kinase activation facilitates epithelial-to-mesenchymal transition in lung cancer by upregulating vimentin expression.
Interleukin 6-triggered ataxia-telangiectasia mutated kinase activation facilitates epithelial-to-mesenchymal transition in lung cancer by upregulating vimentin expression.
Experimental cell research (2019-05-18)
Yi Na Jiang, Xiao Yan Ni, Hong Qiong Yan, Lei Shi, Nan Nan Lu, Yi Nan Wang, Qing Li, Feng Guang Gao
ZUSAMMENFASSUNG
Matrix metalloproteinases (MMPs) and the epithelial-mesenchymal transition (EMT) contribute to metastasis. As shown in our previous studies, interleukin-6 (IL-6) induces ATM phosphorylation to increase MMP expression and metastasis in lung cancer. However, the exact roles of ATM activation in the IL-6-induced epithelial-mesenchymal transition and lung cancer metastasis are currently unclear. Here, ATM phosphorylation exerts its pro-metastatic effect via vimentin-mediated epithelial-mesenchymal transition, which was supported by the evidence described below. Firstly, IL-6 treatment increases vimentin expression via the ATM-NF-κB pathway. Second, ATM inactivation not only abolishes IL-6-induced increases in vimentin expression but also inhibits IL-6-induced nest formation in a xenograft lung metastasis model. Moreover, close positive correlations were observed between ATM phosphorylation and vimentin upregulation, IL-6 levels and metastasis in lung cancer specimens. Hence, ATM modulates vimentin expression to facilitate IL-6-induced epithelial-mesenchymal transition and metastasis in lung cancer, indicating that ATM and vimentin might be potential therapeutic targets for inflammation-associated lung cancer metastasis.