Merck
Genetically modified pigs are protected from classical swine fever virus.
PLoS pathogens (2018-12-14)
Zicong Xie, Daxin Pang, Hongming Yuan, Huping Jiao, Chao Lu, Kankan Wang, Qiangbing Yang, Mengjing Li, Xue Chen, Tingting Yu, Xinrong Chen, Zhen Dai, Yani Peng, Xiaochun Tang, Zhanjun Li, Tiedong Wang, Huancheng Guo, Li Li, Changchun Tu, Liangxue Lai, Hongsheng Ouyang
ZUSAMMENFASSUNG
Classical swine fever (CSF) caused by classical swine fever virus (CSFV) is one of the most detrimental diseases, and leads to significant economic losses in the swine industry. Despite efforts by many government authorities to stamp out the disease from national pig populations, the disease remains widespread. Here, antiviral small hairpin RNAs (shRNAs) were selected and then inserted at the porcine Rosa26 (pRosa26) locus via a CRISPR/Cas9-mediated knock-in strategy. Finally, anti-CSFV transgenic (TG) pigs were produced by somatic nuclear transfer (SCNT). Notably, in vitro and in vivo viral challenge assays further demonstrated that these TG pigs could effectively limit the replication of CSFV and reduce CSFV-associated clinical signs and mortality, and disease resistance could be stably transmitted to the F1-generation. Altogether, our work demonstrated that RNA interference (RNAi) technology combining CRISPR/Cas9 technology offered the possibility to produce TG animal with improved resistance to viral infection. The use of these TG pigs can reduce CSF-related economic losses and this antiviral strategy may be useful for future antiviral research.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Anti-Pig IgG (whole molecule)−FITC antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution