Merck
  • Assembly of Plasmonic and Magnetic Nanoparticles with Fluorescent Silica Shell Layer for Tri-functional SERS-Magnetic-Fluorescence Probes and Its Bioapplications.
Assembly of Plasmonic and Magnetic Nanoparticles with Fluorescent Silica Shell Layer for Tri-functional SERS-Magnetic-Fluorescence Probes and Its Bioapplications.
Scientific reports (2018-09-19)
Hyung-Mo Kim, Dong-Min Kim, Cheolhwan Jeong, So Yeon Park, Myeong Geun Cha, Yuna Ha, Dahye Jang, San Kyeong, Xuan-Hung Pham, Eunil Hahm, Sang Hun Lee, Dae Hong Jeong, Yoon-Sik Lee, Dong-Eun Kim, Bong-Hyun Jun
ZUSAMMENFASSUNG
In this study, we report on the fabrication of multilayered tri-functional magnetic-SERS-fluorescence nanoprobes (MF-SERS particles) containing clustered superparamagnetic Fe3O4 nanoparticles (NPs), silver NPs, and a fluorescent silica layer. The MF-SERS particles exhibited strong SERS signals from the silver NPs as well as both superparamagnetism and fluorescence. MF-SERS particles were uptaken by cells, allowing successful separation using an external magnetic field. SERS and fluorescence signals could be detected from the NP-containing cells, and CD44 antibody-conjugated MF-SERS particles selectively targeted MDA-MB-231 cells. Based on these properties, MF-SERS particles proved to be a useful nanoprobe for multiplex detection and separation of cancer cells.