Merck
  • Experimental polycystic ovarian syndrome is associated with reduced expression and function of P2Y2 receptors in rat theca cells.
Experimental polycystic ovarian syndrome is associated with reduced expression and function of P2Y2 receptors in rat theca cells.
Molecular reproduction and development (2019-01-10)
Anaí Del Rocío Campos-Contreras, Ana Patricia Juárez-Mercado, Adriana González-Gallardo, Rebeca Chávez-Genaro, Edith Garay, Dalia Luz De Ita-Pérez, Mauricio Díaz-Muñoz, Francisco Gabriel Vázquez-Cuevas
ZUSAMMENFASSUNG
Extracellular purines through specific receptors have been recognized as new regulators of ovarian function. It is known that P2Y2 receptor activity induces theca cell proliferation, we hypothesized that purinergic signaling controls the changes related to hyperthecosis in polycystic ovarian syndrome (PCOS). The aim of this study was to analyze the expression of UTP-sensitive P2Y receptors and their role in theca cells (TC) proliferation in experimentally-induced PCOS (EI-PCOS). In primary cultures of TC from intact rats, all the transcripts of P2Y receptors were detected by polymerase chain reaction; in these cells, UTP (10 μM) induced extracellular signal-regulated kinases (ERK) phosphorylation. Rats with EI-PCOS showed a reduced expression of P2Y2R in TC whereas P2Y4R did not change. By analyzing ERK phosphorylation, it was determined that P2Y2R is the most relevant receptor in TC. UTP promoted cell proliferation in TC from control but not from EI-PCOS rats. The in silico analysis of P2yr2 promoter indicated the presence of androgen response elements; the stimulation of TC primary cultures with testosterone promoted a significant reduction in the expression of the P2yr2 transcript. We concluded that P2Y2R participates in controlling the proliferative rate of TCs from healthy ovaries, but this regulation is lost during EI-PCOS.