Merck
Induction of M2 Macrophages Prevents Bone Loss in Murine Periodontitis Models.
Journal of dental research (2018-11-06)
Z Zhuang, S Yoshizawa-Smith, A Glowacki, K Maltos, C Pacheco, M Shehabeldin, M Mulkeen, N Myers, R Chong, K Verdelis, G P Garlet, S Little, C Sfeir
ZUSAMMENFASSUNG
Periodontitis is characterized by the progressive destruction of tooth-supporting alveolar bone, which is mainly caused by chronic inflammation in response to persistent bacterial insult. It has recently become clear that the pathogenesis of periodontitis is associated with a high ratio of proinflammatory M1 (classically activated) macrophages to anti-inflammatory M2 (alternatively activated). To decrease the inflammatory activity, we locally delivered the C-C motif chemokine ligand 2 (CCL2) using controlled-release microparticles (MPs). CCL2 is known to promote chemotaxis of M0 or M2 phenotype macrophages to the inflamed site and induce M2 phenotype polarization locally. Our in vitro data showed that CCL2 increased the number of M2 phenotype macrophages, decreased TNF-α secretion, and enhanced chemotaxis of RAW264.7 cells toward CCL2 MPs. Moreover, we induced periodontal disease in 2 animal models through inoculation of Porphyromonas gingivalis and ligature around the murine molar. Micro-computed tomography analysis showed significant reduction of alveolar bone loss in the CCL2 MP treatment group when compared with a blank MP group and a no-treatment periodontitis group in both models. Immunohistologic analysis showed a significant increase in the M2 phenotype subset and a decrease in the M1 phenotype subset in the CCL2 MP group of the P. gingivalis-induced model. Also, in both models, tartrate-resistant acidic phosphatase staining showed significantly fewer numbers of osteoclasts in the CCL2 MP group in alveolar bone area. Moreover, quantitative polymerase chain reaction results showed a significant increase in IL-1RA (interleukin 1 receptor antagonist) mRNA expression and a decrease in RANKL (receptor activator of nuclear factor kappa-Β ligand) mRNA expression in the CCL2 MP group in the ligature model. In summary, manipulation of endogenous M2 phenotype macrophages with CCL2 MPs decreased the M1 phenotype:M2 phenotype ratio and prevented alveolar bone loss in mouse periodontitis models. The delivery of CCL2 MPs provides a novel approach to treat periodontal disease.