Merck
Phase Inversion in PVDF Films with Enhanced Piezoresponse Through Spin-Coating and Quenching.
Polymers (2019-07-03)
Marco Fortunato, Domenico Cavallini, Giovanni De Bellis, Fabrizio Marra, Alessio Tamburrano, Francesca Sarto, Maria Sabrina Sarto
ZUSAMMENFASSUNG
In the present work, poly(vinylidene fluoride) (PVDF) films were produced by spin-coating, and applying different conditions of quenching, in order to investigate the dominant mechanism of the β-phase formation. The influence of the polymer/solvent mass ratio of the solution, the rotational speed of the spin-coater and the crystallization temperature of the film on both the β-phase content and the piezoelectric coefficient (d33) were investigated. This study demonstrates that the highest values of d33 are obtained when thinner films, produced with a lower concentration of polymer in the solvent (i.e., 20 wt.%), go through quenching in water, at room temperature. Whereas, in the case of higher polymer concentration (i.e., 30 wt.%), the best value of d33 (~30 pm/V) was obtained through quenching in liquid nitrogen, at the temperature of 77 K. We believe that in the former case, phase inversion is mainly originated by electrostatic interaction of PVDF with the polar molecules of water, due to the low viscosity of the polymer solution. On the contrary, in the latter case, due to higher viscosity of the solution, mechanical stretching induced on the polymer during spin-coating deposition is the main factor inducing self-alignment of the β-phase. These findings open up a new way to realize highly efficient devices for energy harvesting and wearable sensors.