Merck
  • Volatile Organic Compound and Fatty Acid Profile of Milk from Cows and Buffaloes Fed Mycorrhizal or Nonmycorrhizal Ensiled Forage.
Volatile Organic Compound and Fatty Acid Profile of Milk from Cows and Buffaloes Fed Mycorrhizal or Nonmycorrhizal Ensiled Forage.
Molecules (Basel, Switzerland) (2019-04-27)
Alessandro Genovese, Andrea Marrazzo, Lucia De Luca, Raffaele Romano, Nadia Manzo, Felicia Masucci, Antonio Di Francia, Raffaele Sacchi
ZUSAMMENFASSUNG
Seed inoculation of forage crops by arbuscular mycorrhizal fungi (AMF) generally results in higher profitability, but also modifies the chemical composition of silage in terms of increased biomass, protein, and dry matter. Raw milk aroma is affected by the type of feed. This work investigated the influence of ensiled forage obtained by seed inoculation with AMF on the volatile fractions and fatty acid composition of milk. Two experiments were carried out: in the first, buffaloes were fed maize silage, and in the second, cows were fed sorghum silage. The volatile fractions of milk were quantified by headspace solid-phase micro-extraction (HS-SPME), combined with gas chromatography-mass spectrometry (GC-MS), and fatty acids by gas chromatography (GC). The ensiled forage obtained with AMF increased saturated fatty acids (SFAs), and decreased monounsaturated fatty acids (MUFAs) and polyunsaturated fatty acids (PUFAs) in both experiments. The volatile fraction in milk samples obtained from bovines fed mycorrhizal ensiled forage showed an increase of free fatty acids and ketones, responsible for cheesy and fruity odors. Aldehydes, responsible for green and grassy notes, increased only in the milk from buffaloes fed ensiled maize. Our results suggest that inoculation of maize and sorghum seed with AMF, combined with a low rate of fertilizers, leads to ensiled forage that could slightly affect the FA profile and odor quality of milk.