Merck
  • NaLa(CO3)2 hybridized with Fe3O4 for efficient phosphate removal: Synthesis and adsorption mechanistic study.
NaLa(CO3)2 hybridized with Fe3O4 for efficient phosphate removal: Synthesis and adsorption mechanistic study.
Water research (2019-03-04)
Haotian Hao, Yili Wang, Baoyou Shi
ZUSAMMENFASSUNG
Effectively eutrophication control and phosphate recovery have received increasing attention in recent years. In this study, a regenerable magnetic NaLa(CO3)2/Fe3O4 composites (MLC) which includes a novel phosphate-binding lanthanum species NaLa(CO3)2 hybridized with Fe3O4 nanoparticle was developed through a modified solvothermal method for phosphate removal from contaminated water. Based upon preliminary screening of synthesized MLC with different La-to-Fe molar ratios in terms of phosphate adsorption capacity and synthetic product yield, a MLC composite with a La-to-Fe molar ratio of 2:1 (MLC-21) was selected for further characterization and evaluation. MLC-21 exhibits a high magnetic separation efficiency of 97%, high phosphate adsorption capacity of 77.85 mg P/g, wide applicable scope of pH ranging from 4 to 11, excellent selectivity for phosphate in the presence of competing ions (Cl-, NO3-, HCO3-, SO42-, Ca2+, and Mg2+), good reusability with above 98% desorption efficiency using NaOHNaCl mixture and 83% adsorption capacity remained during five recycles. Furthermore, a real effluent wastewater with phosphate concentration of 1.96 mg P/L was used to verify the performance of MLC-21 through a magnetic separation integrated system (AMSS). By using the response surface methodology (RSM), the optimum parameters were determined to be 0.26 g/L of adsorbent dosage, 26.28 h of adsorption time and 24.12 min of magnetic separation time for meeting the phosphate emission standard of 0.5 mg P/L. The phosphorus in three representative eutrophic water bodies can be efficiently reduced to below 0.1 mg P/L by MLC-21 adsorption at different dosages. Electrostatic attraction and the inner-sphere complexation between La(HCO3)2+/La(CO3)2- and P via ligand exchange forming LaPO4 were responsible for the phosphate adsorption mechanisms of MLC.