Merck
  • Comparisons of bone density and body composition among adolescents with anorexia nervosa and atypical anorexia nervosa.
Comparisons of bone density and body composition among adolescents with anorexia nervosa and atypical anorexia nervosa.
The International journal of eating disorders (2019-02-17)
Jason M Nagata, Jennifer L Carlson, Neville H Golden, Jin Long, Stuart B Murray, Rebecka Peebles
ZUSAMMENFASSUNG
To compare bone mineral density (BMD) and body composition among adolescents: (a) with atypical anorexia nervosa (AAN) versus anorexia nervosa (AN) and (b) those with and without a prior history of overweight. Electronic medical records of patients 9-20 years with AN or AAN who underwent dual-energy x-ray absorptiometry scans were retrospectively reviewed and analyzed. A total of 286 adolescents with AN or AAN were included. In linear regression models, AAN was associated with greater Z-scores in whole body bone mineral content (BMC, B = 0.88, p < 0.001), lumbar spine BMD (B = 0.79, p = 0.002), femoral neck BMD (B = 0.670, p = 0.009); fat mass index (B = 1.33, p = 0.003), and lean body mass index (LBMI, B = 1.10, p < 0.001) compared to AN, adjusting for age, sex, and duration of illness. A prior overweight history was associated with greater Z-scores in whole body BMC; lumbar spine BMD, total hip BMD, femoral neck BMD, and LBMI. Adolescents with AAN had higher BMD Z-scores than adolescents with AN; adolescents with a prior overweight history had greater BMD Z-scores than adolescents without a prior overweight history. These findings may inform clinical guidelines for the medical management of AAN.