Merck
Eutectic mixture promoted CO2 sorption on MgO-TiO2 composite at elevated temperature.
Journal of environmental sciences (China) (2018-12-12)
Vishwanath Hiremath, Monica Louise T Trivino, Jeong Gil Seo
ZUSAMMENFASSUNG
The development of carbon dioxide (CO2) sorbents that can operate at elevated temperatures is significant for the advancement of pre-combustion capture technologies. Recently, promoter-based systems composed of alkali/alkaline earth metal nitrates and/or carbonates have been considered as next-generation solid sorbents due to their improved CO2 uptake and kinetics. However, obtaining stable MgO sorbents against temperature swing regeneration still remained challenging. Herein, we report MgO-TiO2 solid sorbents promoted by eutectic mixture (KNO3 and LiNO3) for elevated temperature CO2 sorption. The developed sorbents show improved CO2 sorption capacity, which may be attributed to the alternative CO2 sorption pathway provided by the ionization of highly dispersed MgO in the eutectic mixture. The MgO-TiO2 framework was also shown to assist in retaining the MgO configuration by constraining its interaction with CO2. Furthermore, it is demonstrated that constructing composite structures is essential to improve the CO2 sorption characteristics, mainly recyclability, at elevated temperatures. The developed promoter integrated sorbents showed exceptionally high CO2 sorption capacity of >30wt.% at an elevated temperature (300°C) with pronounced stability under temperature swing operation.