Merck
  • Sodium-glucose cotransporter 2 inhibitors regulate ketone body metabolism via inter-organ crosstalk.
Sodium-glucose cotransporter 2 inhibitors regulate ketone body metabolism via inter-organ crosstalk.
Diabetes, obesity & metabolism (2018-11-09)
Jin Hee Kim, Minyoung Lee, Soo Hyun Kim, So Ra Kim, Byung-Wan Lee, Eun Seok Kang, Bong-Soo Cha, Jin Won Cho, Yong-Ho Lee
ZUSAMMENFASSUNG
To investigate sodium-glucose cotransporter 2 inhibitor (SGLT2i)-induced changes in ketogenic enzymes and transporters in normal and diabetic mice models. Normal mice were randomly assigned to receive either vehicle or SGLT2i (25 mg/kg/d by oral gavage) for 7 days. Diabetic mice were treated with vehicle, insulin (4.5 units/kg/d by subcutaneous injection) or SGLT2i (25 mg/kg/d by intra-peritoneal injection) for 5 weeks. Serum and tissues of ketogenic organs were analysed. In both normal and diabetic mice, SGLT2i increased beta-hydroxybutyrate (BHB) content in liver, kidney and colon tissue, as well as in serum and urine. In these organs, SGLT2i upregulated mRNA expression of ketogenic enzymes, 3-hydroxy-3-methylglutaryl-coenzyme A synthase 2 and 3-hydroxy-3-methylglutaryl-coenzyme A lyase. Similar patterns were observed in the kidney, ileum and colon for mRNA and protein expression of sodium-dependent monocarboxylate transporters (SMCTs), which mediate the cellular uptake of BHB and butyrate, an important substrate for intestinal ketogenesis. In diabetic mice under euglycaemic conditions, SGLT2i increased major ketogenic enzymes and SMCTs, while insulin suppressed ketogenesis. SGLT2i increased systemic and tissue BHB levels by upregulating ketogenic enzymes and transporters in the liver, kidney and intestine, suggesting the integrated physiological consequences for ketone body metabolism of SGLT2i administration.