Merck
  • Compound 48/80 reduces the crop-emptying rate, likely through a histamine-associated pathway in chicks.
Compound 48/80 reduces the crop-emptying rate, likely through a histamine-associated pathway in chicks.
Domestic animal endocrinology (2018-11-26)
Tetsuya Tachibana, Wataru Ueoka, Md Sakirul Islam Khan, Ryosuke Makino, Mark A Cline
ZUSAMMENFASSUNG
Infectious conditions are associated with reduced food passage through the digestive tract in both mammals and chicks; however, the precise mechanism mediating this response in chicks remains unclear. The purpose of the present study was to determine if mast cells, a blood cell type which plays an important role in the immune system, might affect food passage through the digestive tract in chicks. Specifically, we performed intraperitoneal (IP) injections of compound 48/80, an inducer of mast cell degranulation, and measured crop emptying. The IP injection of compound 48/80 significantly reduced the crop-emptying rate, but it did not affect the proventriculus to small intestine transit rate or the number of defecations. We also found that IP-injected histamine, which is secreted by mast cells, also reduced the crop-emptying rate. In addition, IP injection of 2-pyridylethylamine (histamine H1 receptor agonist), but not dimaprit, (R)-(-)-α-methylhistamine, and VUF8430 (histamine H2, H3, and H4 receptor agonists, respectively), reduced the crop-emptying rate, implying that histamine reduces the crop emptying rate via the histamine H1 receptor. Finally, we found that IP injection of compound 48/80 reduced mRNA expression of histidine decarboxylase, a rate-limiting enzyme for histamine synthesis, in the esophagus and proventriculus at 1 h and the proventriculus and duodenum at 3 h after the injection. In sum, the present study suggests that the degranulation of mast cells causes a reduction in the crop-emptying rate, possibly via the histamine pathway in chicks.