Merck
  • Cholesteryl Ester Transfer Protein Influences High-Density Lipoprotein Levels and Survival in Sepsis.
Cholesteryl Ester Transfer Protein Influences High-Density Lipoprotein Levels and Survival in Sepsis.
American journal of respiratory and critical care medicine (2018-10-16)
Mark Trinder, Kelly R Genga, HyeJin Julia Kong, Lisanne L Blauw, Cody Lo, Xuan Li, Mihai Cirstea, Yanan Wang, Patrick C N Rensen, James A Russell, Keith R Walley, John H Boyd, Liam R Brunham
ZUSAMMENFASSUNG
High-density lipoprotein (HDL) cholesterol (HDL-C) levels decline during sepsis, and lower levels are associated with worse survival. However, the genetic mechanisms underlying changes in HDL-C during sepsis, and whether the relationship with survival is causative, are largely unknown. We hypothesized that variation in genes involved in HDL metabolism would contribute to changes in HDL-C levels and clinical outcomes during sepsis. We performed targeted resequencing of HDL-related genes in 200 patients admitted to an emergency department with sepsis (Early Infection cohort). We examined the association of genetic variants with HDL-C levels, 28-day survival, 90-day survival, organ dysfunction, and need for vasopressor or ventilatory support. Candidate variants were further assessed in the VASST (Vasopressin versus Norepinephrine Infusion in Patients with Septic Shock Trial) cohort (n = 632) and St. Paul's Hospital Intensive Care Unit 2 (SPHICU2) cohort (n = 203). We identified a rare missense variant in CETP (cholesteryl ester transfer protein gene; rs1800777-A) that was associated with significant reductions in HDL-C levels during sepsis. Carriers of the A allele (n = 10) had decreased survival, more organ failure, and greater need for organ support compared with noncarriers. We replicated this finding in the VASST and SPHICU2 cohorts, in which carriers of rs1800777-A (n = 35 and n = 12, respectively) had significantly reduced 28-day survival. Mendelian randomization was consistent with genetically reduced HDL levels being a causal factor for decreased sepsis survival. Our results identify CETP as a critical regulator of HDL levels and clinical outcomes during sepsis. These data point toward a critical role for HDL in sepsis.