Merck
  • Downregulated miR-621 promotes cell proliferation via targeting CAPRIN1 in hepatocellular carcinoma.
Downregulated miR-621 promotes cell proliferation via targeting CAPRIN1 in hepatocellular carcinoma.
American journal of cancer research (2018-11-13)
Yao Zhang, Wei You, Haoming Zhou, Zhiqiang Chen, Guoyong Han, Xueliang Zuo, Long Zhang, Jindao Wu, Xuehao Wang
ZUSAMMENFASSUNG
MicroRNAs (miRNAs) have been reported to play an essential role in tumor development and progression. However, the function of miR-621 in hepatocellular carcinoma (HCC) remains largely unexplored. In this study, we found that miR-621 was downregulated in the HCC specimens and cell lines, and lower expression of miR-621 indicated poor survival. Overexpression of miR-621 was shown to induce G0/G1 cell cycle arrest and inhibit cell proliferation in vitro and in vivo. Luciferase assays revealed that cell cycle-associated protein 1 (CAPRIN1) is a novel functional downstream target of miR-621. miR-621 could regulate c-MYC and cyclin D2 expression by directly targeting CAPRIN1. Further study revealed that CAPRIN1 was upregulated in the HCC specimens and cell lines. Restoration of CAPRIN1 neutralized the miR-621-induced cell cycle arrest and cell proliferation inhibition. Taken together, our findings suggest that miR-621 acts as a tumor suppressor gene in HCC progression by downregulating CAPRIN1 expression and could be a novel potential diagnostic and prognostic biomarker for HCC.