Merck
  • Halide Ligands To Release Strain in Cadmium Chalcogenide Nanoplatelets and Achieve High Brightness.
Halide Ligands To Release Strain in Cadmium Chalcogenide Nanoplatelets and Achieve High Brightness.
ACS nano (2019-04-13)
Marion Dufour, Junling Qu, Charlie Greboval, Christophe Méthivier, Emmanuel Lhuillier, Sandrine Ithurria
ZUSAMMENFASSUNG
Zinc blende II-VI semiconductor nanoplatelets (NPLs) are defined at the atomic scale along the thickness of the nanoparticle and are initially capped with carboxylates on the top and bottom [001] facets. These ligands are exchanged on CdSe NPLs with halides that act as X-L-type ligands. These CdSe NPLs are costabilized by amines to provide colloidal stability in nonpolar solvents. The hydrogen from the amine can participate in a hydrogen bond with the lone pair electrons of surface halides. After ligand exchange, the optical features are red-shifted. Thus, ligand tuning is another way, in addition to confinement, to tune the optical features of NPLs. The improved surface passivation leads to an increase in the fluorescence quantum efficiency of up to 70% in the case of bromide. However, for chloride and iodide, the surface coverage is incomplete, and thus, the fluorescence quantum efficiency is lower. This ligand exchange is associated with a decrease in stress that leads to unfolding of the NPLs, which is particularly noticeable for iodide-capped NPLs.