Merck
  • Involvement of Src-homology-2-domain-containing protein-tyrosine phosphatase 2 in T cell activation.
Involvement of Src-homology-2-domain-containing protein-tyrosine phosphatase 2 in T cell activation.
European journal of biochemistry (1996-05-01)
P Tailor, T Jascur, S Williams, M von Willebrand, C Couture, T Mustelin
ZUSAMMENFASSUNG
Activation of resting T lymphocytes by ligands to the complex of T cell antigen receptor (TCR) and CD3 is initiated by a series of critical tyrosine phosphorylation and dephosphorylation events. Protein-tyrosine kinases of the Syk, Src and Csk families and the CD45 protein-tyrosine phosphatase (PTPase) are known to be involved in these early biochemical reactions. We have found that one of the two T-cell-expressed SH2-domain-containing PTPases, SHPTP2, is rapidly phosphorylated on tyrosine upon addition of anti-CD3 mAbs. This response was absent in cells lacking the Src family kinase Lck. Concomitantly with tyrosine phosphorylation, SHPTP2 co-immunoprecipitated with two unphosphorylated cellular proteins; phosphatidylinositol 3-kinase p85 and Grb2. Binding of SHPTP2 to Grb2 occurred through the SH2 domain of Grb2, while the association between SHPTP2 and p85 seemed to be mediated through Grb2 as an intermediate. In addition, many other molecules associate with Grb2 and may thereby become juxtaposed to SHPTP2. Our results indicate that SHPTP2 participates actively at an early stage in TCR signaling and that its phosphorylation on tyrosine may direct a Grb2-dependent association with selected substrates.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
PTP Assay Kit 1, PTP Assay Kit 1 can be used to detect PTP-1B activity by dephosphorylation of the phosphopeptide (RRLIEDAEpYAARG) using malachite green detection, or by hydrolysis of pNPP.