- Nuclease activities in a complex of human recombination and DNA repair factors Rad50, Mre11, and p95.
Nuclease activities in a complex of human recombination and DNA repair factors Rad50, Mre11, and p95.
The Journal of biological chemistry (1998-08-15)
K M Trujillo, S S Yuan, E Y Lee, P Sung
PMID9705271
ZUSAMMENFASSUNG
Genetic studies in yeast have indicated a role of the RAD50 and MRE11 genes in homologous recombination, telomere length maintenance, and DNA repair processes. Here, we purify from nuclear extract of Raji cells a complex consisting of human Rad50, Mre11, and another protein factor with a size of about 95 kDa (p95), which is likely to be Nibrin, the protein encoded by the gene mutated in Nijmegen breakage syndrome. We show that the Rad50-Mre11-p95 complex possesses manganese-dependent single-stranded DNA endonuclease and 3' to 5' exonuclease activities. These nuclease activities are likely to be important for recombination, repair, and genomic stability.