Merck
Glutamate-dependent active-site labeling of brain glutamate decarboxylase.
Journal of neurochemistry (1990-08-01)
D L Martin, S J Wu, S B Martin
ZUSAMMENFASSUNG
A major regulatory feature of brain glutamate decarboxylase (GAD) is a cyclic reaction that controls the relative amounts of holoenzyme and apoenzyme [active and inactive GAD with and without bound pyridoxal 5'-phosphate (pyridoxal-P, the cofactor), respectively]. Previous studies have indicated that progression of the enzyme around the cycle should be stimulated strongly by the substrate, glutamate. To test this prediction, the effect of glutamate on the incorporation of pyridoxal-P into rat-brain GAD was studied by incubating GAD with [32P]pyridoxal-P, followed by reduction with NaBH4 to link irreversibly the cofactor to the enzyme. Adding glutamate to the reaction mixture strongly stimulated labeling of GAD, as expected. 4-Deoxypyridoxine 5'-phosphate (deoxypyridoxine-P), a close structural analogue of pyridoxal-P, was a competitive inhibitor of the activation of glutamate apodecarboxylase by pyridoxal-P (Ki = 0.27 microM) and strongly inhibited glutamate-dependent labeling of GAD. Analysis of labeled GAD by sodium dodecyl sulfate (SDS)-polyacrylamide gel electrophoresis showed two labeled proteins with apparent molecular masses of 59 and 63 kDa. Both proteins could be purified by immunoaffinity chromatography on a column prepared with a monoclonal antibody to GAD, and both were labeled in a glutamate-dependent, deoxypyridoxine-P-sensitive manner, indicating that both were GAD. Three peaks of GAD activity (termed peaks I, II, and III) were separated by chromatography on phenyl-Sepharose, labeled with [32P]pyridoxal-P, purified by immunoaffinity chromatography, and analyzed by SDS-polyacrylamide gel electrophoresis. Peak I contained only the 59-kDa labeled protein. Peaks II and III contained the both the 59- and 63-kDa proteins, but in differing proportions.(ABSTRACT TRUNCATED AT 250 WORDS)
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Adenosin-5′-Monophosphat-Agarose, lyophilized powder