Merck
Abnormal vascular function and hypertension in mice deficient in estrogen receptor beta.
Science (New York, N.Y.) (2002-01-19)
Yan Zhu, Zhao Bian, Ping Lu, Richard H Karas, Lin Bao, Daniel Cox, Jeffrey Hodgin, Philip W Shaul, Peter Thoren, Oliver Smithies, Jan-Ake Gustafsson, Michael E Mendelsohn
ZUSAMMENFASSUNG
Blood vessels express estrogen receptors, but their role in cardiovascular physiology is not well understood. We show that vascular smooth muscle cells and blood vessels from estrogen receptor beta (ERbeta)-deficient mice exhibit multiple functional abnormalities. In wild-type mouse blood vessels, estrogen attenuates vasoconstriction by an ERbeta-mediated increase in inducible nitric oxide synthase expression. In contrast, estrogen augments vasoconstriction in blood vessels from ERbeta-deficient mice. Vascular smooth muscle cells isolated from ERbeta-deficient mice show multiple abnormalities of ion channel function. Furthermore, ERbeta-deficient mice develop sustained systolic and diastolic hypertension as they age. These data support an essential role for ERbeta in the regulation of vascular function and blood pressure.