Merck
Role of cardiac glycosides in regulation of the growth of retinal tissue explants.
Bulletin of experimental biology and medicine (2009-06-11)
E V Lopatina, A V Karetsky, V A Penniyaynen, T V Vinogradova
ZUSAMMENFASSUNG
We studied the role of cardiac glycosides in the regulation of the growth of retinal tissue explants from 10-12-day-old chicken embryos in organotypic culture. The studied compounds produced a dose-dependent effect on cell proliferation in retinal tissue explants. Ouabain (10(-13)M), strophanthin K (10(-13)M) and digoxin (10(-11)M) significantly stimulated explant growth. It was hypothesized that the physiological role of endogenous ouabain is associated with regulation of tissue modeling.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Cymarin, ≥96% (HPLC)