Merck
  • Tumor-inhibitory antibodies to HER-2/ErbB-2 may act by recruiting c-Cbl and enhancing ubiquitination of HER-2.
Tumor-inhibitory antibodies to HER-2/ErbB-2 may act by recruiting c-Cbl and enhancing ubiquitination of HER-2.
Cancer research (2000-07-26)
L N Klapper, H Waterman, M Sela, Y Yarden
ZUSAMMENFASSUNG
Overexpression of HER-2/ErbB-2, a homologue of the epidermal growth factor receptor, is associated with poor prognosis, and an ErbB-2-specific antibody is therapeutic when administered to patients with metastatic breast cancer. To understand the mechanism underlying immunotherapy, we concentrated on antibody- and epidermal growth factor-induced degradation of ErbB-2. We show that enhanced degradation is preceded by poly-ubiquitination of ErbB-2. This process necessitates recruitment of the c-Cbl ubiquitin ligase to tyrosine 1112 of ErbB-2. Consequently, mutagenesis of this site retards antibody-induced degradation. Thus, the therapeutic potential of certain antibodies may be due to their ability to direct ErbB-2 to a c-Cbl-regulated proteolytic pathway.