Merck
Palladium(III)-catalyzed fluorination of arylboronic acid derivatives.
Journal of the American Chemical Society (2013-09-18)
Anthony R Mazzotti, Michael G Campbell, Pingping Tang, Jennifer M Murphy, Tobias Ritter
ZUSAMMENFASSUNG
A practical, palladium-catalyzed synthesis of aryl fluorides from arylboronic acid derivatives is presented. The reaction is operationally simple and amenable to multigram-scale synthesis. Evaluation of the reaction mechanism suggests a single-electron-transfer pathway, involving a Pd(III) intermediate that has been isolated and characterized.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
[Pd(terpy)(MeCN)][BF4]2, ≥95%