Merck
Complement 7 Is Up-Regulated in Human Early Diabetic Kidney Disease.
The American journal of pathology (2018-09-27)
Monica Sircar, Ivy A Rosales, Martin K Selig, Dihua Xu, Zsuzsanna K Zsengeller, Isaac E Stillman, Towia A Libermann, S Ananth Karumanchi, Ravi I Thadhani
ZUSAMMENFASSUNG
There is a temporal window from the time diabetes is diagnosed to the appearance of overt kidney disease during which time the disease progresses quietly without detection. Currently, there is no way to detect early diabetic nephropathy (EDN). Herein, we performed an unbiased assessment of gene-expression analysis of postmortem human kidneys to identify candidate genes that may contribute to EDN. We then studied one of the most promising differentially expressed genes in both kidney tissue and blood samples. Differential transcriptome analysis of EDN kidneys and matched nondiabetic controls showed alterations in five canonical pathways, and among them the complement pathway was the most significantly altered. One specific complement pathway gene, complement 7 (C7), was significantly elevated in EDN kidney. Real-time PCR confirmed more than a twofold increase of C7 expression in EDN kidneys compared with controls. Changes in C7 gene product level were confirmed by immunohistochemistry. C7 protein levels were elevated in proximal tubules of EDN kidneys. Serum C7 protein levels were also measured in EDN and control donors. C7 levels were significantly higher in EDN serum than control serum. This latter finding was independently confirmed in a second set of blood samples from a previously collected data set. Together, our data suggest that C7 is associated with EDN, and can be used as a molecular target for detection and/or treatment of EDN.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Anti-C7 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution