Merck
  • Design and synthesis of a stable oxidized phospholipid mimic with specific binding recognition for macrophage scavenger receptors.
Design and synthesis of a stable oxidized phospholipid mimic with specific binding recognition for macrophage scavenger receptors.
Journal of medicinal chemistry (2012-09-01)
William W Turner, Karsten Hartvigsen, Agnes Boullier, Erica N Montano, Joseph L Witztum, Michael S VanNieuwenhze
ZUSAMMENFASSUNG
Macrophage scavenger receptors appear to play a major role in the clearance of oxidized phospholipid (OxPL) products. Discrete peptide-phospholipid conjugates with the phosphatidylcholine headgroup have been shown to exhibit binding affinity for these receptors. We report the preparation of a water-soluble, stable peptide-phospholipid conjugate (9) that possesses the necessary physical properties to enable more detailed study of the role(s) of OxPL in metabolic disease.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Avanti
16:0-20:4 PC, Avanti Research - A Croda Brand